Learn More
Ethos Biosciences AFB Kinyoun Carbol Fuchsin
Catalog No. 5000819179
Encompass_Preferred
AFB Kinyoun Carbol Fuchsin is formulated for use as the primary stain in acid-fast staining procedures including the Kinyoun cold method It penetrates the waxy cell walls of acid-fast organisms producing a strong red coloration that resists decolorization with acid-alcohol This allows clear differentiation of acid-fast bacteria from non acid-fast material after application of a suitable counterstain Suitable for routine use in clinical veterinary and research laboratories the reagent is compatible with standard acid-fast staining protocols and manual procedures The formulation supports consistent staining performance and dependable results in microbiology applications
Catalog No. 50-008-19179 Supplier Ethos Biosciences Supplier No. 630108
AFB Kinyoun Carbol Fuchsin is formulated for use as the primary stain in acid-fast staining procedures including the Kinyoun cold method It penetrates the waxy cell walls of acid-fast organisms producing a strong red coloration that resists decolorization with acid-alcohol This allows clear differentiation of acid-fast bacteria from non acid-fast material after application of a suitable counterstain Suitable for routine use in clinical veterinary and research laboratories the reagent is compatible with standard acid-fast staining protocols and manual procedures The formulation supports consistent staining performance and dependable results in microbiology applications