Learn More
Ethos Biosciences AFB Kinyoun Methylene Blue
Catalog No. 5000819196
Encompass_Preferred
AFB Methylene Blue Counterstain is formulated for use as a counterstain in acid-fast staining procedures including the Kinyoun and Ziehl-Neelsen methods It provides a clear blue background that enhances contrast with the red-stained acid-fast organisms supporting accurate microscopic differentiation Designed for routine use in clinical veterinary and research laboratories this counterstain is compatible with standard acid-fast staining protocols and manual procedures The formulation delivers consistent color development and reliable performance for dependable staining results in microbiology applications
Catalog No. 50-008-19196 Supplier Ethos Biosciences Supplier No. 630508
AFB Methylene Blue Counterstain is formulated for use as a counterstain in acid-fast staining procedures including the Kinyoun and Ziehl-Neelsen methods It provides a clear blue background that enhances contrast with the red-stained acid-fast organisms supporting accurate microscopic differentiation Designed for routine use in clinical veterinary and research laboratories this counterstain is compatible with standard acid-fast staining protocols and manual procedures The formulation delivers consistent color development and reliable performance for dependable staining results in microbiology applications