Learn More
IMEB, Inc AFB Stain Kit, Includes: 1- 8 Oz. Bottle/Solutions of: Fuchsin Stain, Phenol Dilute, Decolorizing, 1 Light Green SF Counterstain, 4- 2 Oz. Staining Containers. and Storage Tray.
SDP
Catalog No. 501689472
Encompass_Preferred
Acid Fast Bacteria Stain using Light Green SF for counterstain. The method provides a visualization of red bacteria against a green background of remaining tissue components. Includes: 1- 8 oz. Bottle of Fuchsin Stain solution, 1- 8 oz. Bottle of Phenol dilute solution, 1- 8 oz. Bottle Decolorizing solution, 1- 8 oz. Bottle Light Green SF Counterstain, 4- 2 oz. Staining containers and storage tray
Catalog No. 50-168-9472 Supplier IMEB, Inc Supplier No. K7008
Acid Fast Bacteria Stain using Light Green SF for counterstain. The method provides a visualization of red bacteria against a green background of remaining tissue components. Includes: 1- 8 oz. Bottle of Fuchsin Stain solution, 1- 8 oz. Bottle of Phenol dilute solution, 1- 8 oz. Bottle Decolorizing solution, 1- 8 oz. Bottle Light Green SF Counterstain, 4- 2 oz. Staining containers and storage tray