Learn More
Newcomer Supply AFB, Fite Stain Kit, 250 ml kit
SDP
Catalog No. NC0779518
Encompass
Newcomer Supply AFB, Fite Stain Kit procedure is used to detect the presence of either Nocardia sp. or Mycobacterium leprae sp. (causative agent of leprosy) in tissue sections. Procedural variations are included for detection of either organism.
Catalog No. NC0779518 Supplier Newcomer Supply Supplier No. 91013A
Newcomer Supply AFB, Fite Stain Kit procedure is used to detect the presence of either Nocardia sp. or Mycobacterium leprae sp. (causative agent of leprosy) in tissue sections. Procedural variations are included for detection of either organism.