Learn More
Newcomer Supply AFB, Ziehl-Neelsen Stain Kit, 250 ml kit
SDP
Catalog No. NC9527267
Encompass
Newcomer Supply AFB, Ziehl-Neelsen Stain Kit procedure is used to demonstrate the presence of acid-fast mycobacteria in tissue sections. Carbol Fuchsin Stain, Ziehl-Neelson, combines phenol and basic fuchsin that works to permeate the lipoid capsule of acid-fast organisms and render them resistant to acid alcohol decolorization.
Catalog No. NC9527267 Supplier Newcomer Supply Supplier No. 9101A
Newcomer Supply AFB, Ziehl-Neelsen Stain Kit procedure is used to demonstrate the presence of acid-fast mycobacteria in tissue sections. Carbol Fuchsin Stain, Ziehl-Neelson, combines phenol and basic fuchsin that works to permeate the lipoid capsule of acid-fast organisms and render them resistant to acid alcohol decolorization.