Learn More
Abcam AFC (7-Amino-4-trifluoromethylcoumarin), Fluorescent marker
Catalog No. NC2680318
Encompass_Preferred
Fluorescent marker. Substrate for fluorogenic enzyme assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2680318 Supplier Abcam Supplier No. AB23039550UL
Fluorescent marker. Substrate for fluorogenic enzyme assays.
The product is subject to the following: Abcam Restricted Use Statement