Learn More
AdipoGen AFC Standard Solution
SDP
Catalog No. 502562165
Encompass_Preferred
Chemical. CAS 53518-15-3. Formula C10H6F3NO2. MW 229.2. A fluorogenic standard useful for quantitating assays monitoring 7-amino-4-trifluoromethylcoumarin AFC release such as those utilizing ubiquitin-AFC. The concentration of the AFC released can be calculated from a standard curve prepared with AFC standards.
Catalog No. 50-256-2165 Supplier AdipoGen Supplier No. AGCS10104R100
Chemical. CAS 53518-15-3. Formula C10H6F3NO2. MW 229.2. A fluorogenic standard useful for quantitating assays monitoring 7-amino-4-trifluoromethylcoumarin AFC release such as those utilizing ubiquitin-AFC. The concentration of the AFC released can be calculated from a standard curve prepared with AFC standards.