Learn More
New England Biolabs, Inc. AfeI – 1000 units
SDP
Catalog No. 50994891
Encompass
A restriction endonuclease that recognizes the sequence AGC_^GCT.
Catalog No. 50-994-891 Supplier New England Biolabs, Inc. Supplier No. R0652L
A restriction endonuclease that recognizes the sequence AGC_^GCT.