Learn More
Cayman Chemical aFetoproteIn MoncLNal ntI
SDP
Catalog No. 502916333
Encompass_Preferred
Immunogen: Recombinant human α-fetoprotein • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) α-Fetoprotein • Applications: FC
Catalog No. 50-291-6333 Supplier Cayman Chemical Supplier No. 38100100 TEST
Immunogen: Recombinant human α-fetoprotein • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) α-Fetoprotein • Applications: FC