Learn More
Aviva Systems Biology Corp AFF2 Antibody - N-terminal region (ARP34260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AFF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AFF2. Alternate names: FMR2, FRAXE, MRX2, OX19, FMR2PApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55314