Learn More
Bio Rad Laboratories AFFI GEL 10 GEL 25ML
Catalog No. NC9331627
Encompass
Affi-Gel 10 Gel, 25mL, provides spontaneous, rapid, and lighly efficient coupling of ligands via primary amines
Catalog No. NC9331627 Supplier Bio Rad Laboratories Supplier No. 1536099
Affi-Gel 10 Gel, 25mL, provides spontaneous, rapid, and lighly efficient coupling of ligands via primary amines