Learn More
Jackson Immuno Research Labs AFFINIPURE DONKEY ANTI-MOUSE
SDP
Supplier:  Jackson Immuno Research Labs 715675150
Encompass_Preferred
Brilliant Violet 421™ AffiniPure Donkey Anti-Mouse IgG (H+L)(min X Bov, Ck, Gt, GP, Sy Hms, Hrs, Hu, Rb, Shp Sr Prot)
Catalog No. NC1505193