Learn More
Jackson Immuno Research Labs AffiniPure Goat Anti-Alpaca IgG, subclasses 2+3 specific | 0.5mg
SDP
Supplier:  Jackson Immuno Research Labs 128005229
Encompass_Preferred
Target: Alpaca | Host: Goat | Antibody Format: Whole IgG | Specificity: IgG, subclasses 2+3 specific | Minimal Cross Reactivity: Bovine, Human, Mouse, Rabbit, Rat Serum Proteins | Conjugate: Unconjugated
Catalog No. NC1681436
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov