Learn More
Jackson Immuno Research Labs AffiniPure Goat Anti-Alpaca IgG, VHH domain | 0.25mg
SDP
Catalog No. NC1681439
Encompass_Preferred
Target: Alpaca | Host: Goat | Antibody Format: Whole IgG | Specificity: IgG, VHH domain | Minimal Cross Reactivity: Bovine, Human, Mouse, Rabbit, Rat Serum Proteins | Conjugate: Unconjugated
Catalog No. NC1681439 Supplier Jackson Immuno Research Labs Supplier No. 128005230
Target: Alpaca | Host: Goat | Antibody Format: Whole IgG | Specificity: IgG, VHH domain | Minimal Cross Reactivity: Bovine, Human, Mouse, Rabbit, Rat Serum Proteins | Conjugate: Unconjugated