Learn More
Jackson Immuno Research Labs AffiniPure Goat Anti-Cat IgG, Fc fragment specific | 2.0mg
SDP
Supplier:  Jackson Immuno Research Labs 102005008
Encompass_Preferred
Target: Cat | Host: Goat | Antibody Format: Whole IgG | Specificity: IgG, Fc fragment specific | Conjugate: Unconjugated
Catalog No. NC1257490