Learn More
World Precision Instrument Affordable High Performance Peristaltic Pump-PERIPRO-8LS
SDP
Catalog No. 502538636
Encompass
Peri-Star Pro peristaltic pumps provide accurate and precise pumping with convenience and versatility. Peri-Star Pro can be run in either flow rate mode (mL/min) or rotation speed mode (rpm). For good laboratory practice, pumps must be calibrated after changing the tubing and solution. You can easily calibrate Peri-Star Pro to deliver flow as accurate as 0.5% in a wide flow range from 0.1 - 280 mL/min. Under rotation speed mode, the digitally controlled stepping motor provides accurate and reproducible operation with 0.1% rpm both forward and in reverse.
Catalog No. 50-253-8636 Supplier World Precision Instrument Supplier No. PERIPRO8LS
Peri-Star Pro peristaltic pumps provide accurate and precise pumping with convenience and versatility. Peri-Star Pro can be run in either flow rate mode (mL/min) or rotation speed mode (rpm). For good laboratory practice, pumps must be calibrated after changing the tubing and solution. You can easily calibrate Peri-Star Pro to deliver flow as accurate as 0.5% in a wide flow range from 0.1 - 280 mL/min. Under rotation speed mode, the digitally controlled stepping motor provides accurate and reproducible operation with 0.1% rpm both forward and in reverse.