Learn More
Aviva Systems Biology Corp AFG3L2 Antibody - middle region (ARP46780_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AFG3L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AFG3L2. Alternate names: FLJ25993, SCA28ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59397