Learn More
ABclonal Technology AFG3L2 Rabbit pAb
SDP
Catalog No. 5000106069
Encompass_Preferred
This gene encodes a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. This gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.
Catalog No. 50-001-06069 Supplier ABclonal Technology Supplier No. A15393500UL
This gene encodes a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. This gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.