Learn More
Medchemexpress LLC Afimetoran (BMS-986256) | 2171019-55-7 | 99.4% | 444.57 | 50 MG
SDP
Catalog No. 5000348924
Encompass_Preferred
Afimetoran (BMS-986256) is an orally active, selective, and highly bioavailable antagonist of Toll-like Receptor 7 and 8 (TLR7/8). It operates by inhibiting the activation of the NF-κB pathway, which is crucial in immune responses. Furthermore, Afimetoran can reverse TLR7-mediated resistance to steroid-induced apoptosis in plasma cell-like dendritic cells (pDCs). This compound is valuable for research into inflammation and autoimmune diseases, such as systemic lupus erythematosus.
  • Orally active
  • Selective TLR7/8 antagonist
  • Highly bioavailable
  • Inhibits NF-κB pathway
  • Reverses steroid resistance
  • Research applications
  • Demonstrated in vivo efficacy
Catalog No. 50-003-48924 Supplier Medchemexpress LLC Supplier No. HY13956750MG
Afimetoran (BMS-986256) is an orally active, selective, and highly bioavailable antagonist of Toll-like Receptor 7 and 8 (TLR7/8). It operates by inhibiting the activation of the NF-κB pathway, which is crucial in immune responses. Furthermore, Afimetoran can reverse TLR7-mediated resistance to steroid-induced apoptosis in plasma cell-like dendritic cells (pDCs). This compound is valuable for research into inflammation and autoimmune diseases, such as systemic lupus erythematosus.
  • Orally active
  • Selective TLR7/8 antagonist
  • Highly bioavailable
  • Inhibits NF-κB pathway
  • Reverses steroid resistance
  • Research applications
  • Demonstrated in vivo efficacy