Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion™ 2 CLIA Moderate Complexity Analyzer Rx

Catalog No. 23050100 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Sample Type:
Whole Blood, Plasma, or Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
23-050-100 Whole Blood, Plasma, or Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-050-100 Supplier Abbott™ Supplier No. 1116970
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Afinion™ 2 Analyzer is a compact, rapid, multi-assay analyzer that provides valuable near patient testing at the point-of-care

  • Fast and easy quantitative determinations of Hemoglobin A1c (HbA1c) and Albumin-Creatinine Ratio (ACR)
  • Four tests on one platform; HbA1c, Lipid Panel, ACR, CRP
  • CLIA Classification: CLIA Moderate Complexity
  • Factory calibrated - no calibration check devices or costly operator calibration is required
  • No maintenance - analyzer does not contain parts that require periodic replacement
  • Extensive and comprehensive failsafe systems prevent the possibility of erroneous results
  • Test results can be printed or transferred to electronic medical records
  • Compact size and short test times
  • Guided test procedure

Specifications

Type Point-of-Care Analyzer
Sample Type Whole Blood, Plasma, or Urine
For Use With (Application) Quantitative Determination of HbA1c and ACR
Includes Afinion™ 2 Analyzer, Quick Reference Guide, User Manual, Power Cord
Dimensions (D x W x H) 7.2 x 7.5 x 13 in.
Quality Control Details QC and Operator Lockout
Weight (English) 7.5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.