Learn More
Abbott™ Afinion™ 2 CLIA Waived Analyzer Rx
Catalog No. 23050111 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Sample Type:
Whole Blood, Urine
Catalog No. Sample Type
23-050-111 Whole Blood, Urine
1 options
This item is not returnable. View return policy
Catalog No. 23-050-111 Supplier Abbott™ Supplier No. 1116985
Add to Cart
Add to Cart
This item is not returnable. View return policy
A compact, rapid, multi-assay analyzer that provides valuable near patient testing at the point-of-care
  • Fast and easy quantitative determinations of Hemoglobin A1c (HbA1c) and Albumin-Creatinine Ratio (ACR)
  • Four tests on one platform; HbA1c, Lipid Panel, ACR, CRP
  • CLIA Classification: CLIA Waived
  • Factory calibrated - no calibration check devices or costly operator calibration is required
  • No maintenance - analyzer does not contain parts that require periodic replacement
  • Extensive and comprehensive failsafe systems prevent the possibility of erroneous results
  • Test results can be printed or transferred to electronic medical records
  • Compact size and short test times
  • Guided test procedure
Specifications
Type Point-of-Care Analyzer
Sample Type Whole Blood, Urine
Description CLIA Waived
Includes Affinion™ 2 Analyzer, Quick Reference Guide, User Manual, Power Guide
Dimensions (D x W x H) 7.2 x 7.5 x 13 in.
Weight (English) 7.5 lb.