Learn More
Abbott™ Afinion™ ACR Control
Catalog No. 23050103 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
(2) 1 mL/Pk.
Catalog No. Quantity
23-050-103 (2) 1 mL/Pk.
1 options
Catalog No. 23-050-103 Supplier Abbott™ Supplier No. 1116973
Add to Cart
Edge
Add to Cart
Afinion™ ACR Control contains liquid preparations of albumin and creatinine in citrate buffer; designed for use with the Afinion System
  • 3.5 μL random/spot urine sample
  • Higly accurate results in 5 minutes
  • Increased accuracy by measuring creatinine
  • Quality control and operator lockout
  • No sample preparation
  • Measuring Range
    • Albumin: 5.0 - 200.0 mg/L
    • Creatinine: 16.4 - 339.9 mg/dL (1.5 - 30.0 mmol/L)
    • ACR: 1.0 - 1225.0 mg/g (0.1 - 140.0 mg/mmol)
Specifications
For Use With (Equipment) Afinion™ System
Type ACR Control
Quantity (2) 1 mL/Pk.
Time Range 5 min.