Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion™ 2 Analyzer, Tests and Controls Kit

Catalog No. 23046653 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
No. of Tests:
15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
23-046-653 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Order Info: Order eligibility is based on upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.

Catalog No. 23-046-653 Supplier Abbott™ Supplier No. 1115175MPA
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Order Info: Order eligibility is based on upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.

Includes

1116554 (Analyzer), 1115015 (HbA1c Test Cartridges) and 1115178 (HbA1c Controls)

Afinion™ Analyzer, Tests and Controls Kit includes Products 1116554 (Analyzer), 1115015 (HbA1c Test Cartridges) and 1115178 (HbA1c Controls) for an all-inclusive on-the-spot testing, regardless of the sample type to deliver accurate results during patient consultation

Afinion 2 Analyzer:

Afinion 2 system is ideal for point-of-care testing in physician offices, clinics, community health centers and hospital out-patient clinics. From just a small urine or fingerstick whole blood sample, lab-accurate results for ACR and HbA1c are made available during the consultation.

Reduced workload in the practice:

  • Fewer follow-up calls
  • Fewer follow-up letters
  • Fewer patient revisits
  • More informed decision-making improves patientclinician relationships, patient motivation and attitude

HbA1c Test Cartridge:

Gives reliable results conveniently. Patient consultations can be carried out with confidence. The HbA1c Test Cartridge contains all reagents necessary for the measurement of glycated hemoglobin.

  • Used to monitor metabolic control in patients with diabetes
  • Aids in diagnosis of diabetes and identifying patients at risk for developing diabetes
  • Sample volume: 1.5 μL
  • Assay time: 3 minutes
  • Measuring range: 4 - 15% HbA1c, 20-140 mmol/mol
  • Hb-range: 6 - 20 g/dL
  • Total imprecision, external studies: < 3% CV
  • Traceable to the IFCC Reference Method
  • Reports DCCT / UKPDS aligned values
  • No influence from Hb variants like HbC, HbD, HbE, HbF, HbJ and HbS
  • No influence from carbamylated or pre-glycated hemoglobin
  • Kit size: 15 tests. Separately packed cartridges
  • Afinion™ HbA1c Control: 2 levels, ready to use
  • Sample material: Capillary blood or anticoagulated venous blood (EDTA, citrate or heparin)
  • Total imprecision: <3% CV
  • Store test kit refrigerated (2° to 8°C) until expiration date or at room temperature for a maximum of 90 days

HbA1c Control:

For use with the Afinion™ AS100 Analyzer and the Afinion™ HbA1c test

  • Contains liquid preparations of stabilized porcine whole blood (Control C I) and human whole blood (Control C II)
  • Quality control testing should be done to confirm that your Afinion™ Analyzer System is working properly and provides reliable results
  • Only when controls are used routinely and the values obtained are within acceptable ranges can accurate esults for patient samples be assured

Order Info

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.

Specifications

Type Point-of-Care Analyzer
Sample Type Capillary and Venous Blood
For Use With (Application) Used to monitor metabolic control in patients with diabetes
Analysis Time 3 min.
No. of Tests 15
Sample Size 1.5 μL
Includes Analyzer, HbA1c Test Cartridges and HbA1c Controls
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.