Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion™ HbA1c Calibration Verification

Catalog No. 23050110 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
3 Levels/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-050-110 3 Levels/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-050-110 Supplier Abbott™ Supplier No. 1116792
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Use with the Alere Afinion™ AS100 Analyzer/Afinion™ 2 Analyzer System to verify the calibration of the Afinion™ HbA1c Dx Test throughout the measurable range

  • Ready to use controls and calibration verification materials
  • No interference from common Hb variants
  • Fingerstick and venous whole blood samples
  • Sample Size: 1.5 μL
  • NGSP and IFCC Certified
  • Factory calibrated instrument – no user calibration required
  • Storage: Up to 3 months at room temperature
  • Test time: 3 minutes

Specifications

Certifications/Compliance NGSP and IFCC Certified
Content And Storage Up to 3 months (at Room Temperature)
Description Ready to Use Controls and Calibration Verification Materials
For Use With (Equipment) Alere Afinion™ AS100 Analyzer, Afinion™ 2 Analyzer System
Quantity 3 Levels/Pk.
Time Range 3 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.