Learn More
Medchemexpress LLC HY-120051 10mg Medchemexpress, Afizagabar CAS:1398496-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12005110MG
Encompass_Preferred
Medchemexpress, HY-120051 10mg Afizagabar CAS:1398496-82-6 Afizagabar (S44819) is a first-in-class, competitive, and selective antagonist at the GABA-binding site of the α5-GABAAR, with an IC50 of 585 nM for α5β2γ2 and a Ki of 66 nM for α5β3γ2. Afizagabar enhances hippocampal synaptic plasticity and exhibits pro-cognitive efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0934