Learn More
Medchemexpress LLC Afizagabar | 1398496-82-6 | 98.4% | 5 MG
SDP
Catalog No. 5000233480
Encompass_Preferred
Afizagabar is a competitive and selective antagonist of the α5 subtype of the GABA-A receptor (α5-GABAAR), used in research to probe GABAergic signaling and cognitive function. It is supplied as a high-purity solid or as a 10 mM solution in DMSO for laboratory use.
  • Competitive and selective α5-GABAAR antagonist.
  • Enhances hippocampal synaptic plasticity and shows pro-cognitive effects in models.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats.
  • Recommended storage: powder -20°C long term; solutions at -80°C.
Catalog No. 50-002-33480 Supplier Medchemexpress LLC Supplier No. HY1200515MG
Afizagabar is a competitive and selective antagonist of the α5 subtype of the GABA-A receptor (α5-GABAAR), used in research to probe GABAergic signaling and cognitive function. It is supplied as a high-purity solid or as a 10 mM solution in DMSO for laboratory use.
  • Competitive and selective α5-GABAAR antagonist.
  • Enhances hippocampal synaptic plasticity and shows pro-cognitive effects in models.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats.
  • Recommended storage: powder -20°C long term; solutions at -80°C.