Learn More
Enzo Life Sciences Aflatoxicol (1mg). CAS: 29611-03-8
SDP
Catalog No. 502013378
Encompass_Preferred
Rare Aflatoxin that is similar but less toxic than Aflatoxin B1. Exposure may produce necrosis, cirrhosis and carcinoma of the liver. Induces DNA damage. In vitro metabolite of Aflatoxin B1. Alternative name: Aflatoxin R0. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO, ethyl acetate, methanol, ethanol, or petrol ether. Not soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-3378 Supplier Enzo Life Sciences Supplier No. ENZCHM1040001
Rare Aflatoxin that is similar but less toxic than Aflatoxin B1. Exposure may produce necrosis, cirrhosis and carcinoma of the liver. Induces DNA damage. In vitro metabolite of Aflatoxin B1. Alternative name: Aflatoxin R0. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO, ethyl acetate, methanol, ethanol, or petrol ether. Not soluble in water. Long Term Storage: -20°C.