Learn More
Medchemexpress LLC HY-N6615 1mg Medchemexpress, Aflatoxin B1 CAS:1162-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66151MG
Encompass_Preferred
Medchemexpress, HY-N6615 1mg Aflatoxin B1 CAS:1162-65-8 Aflatoxin B1 (AFB1) is a Class 1A carcinogen, which is a secondary metabolite of Aspergillus flavus and A. parasiticus. Aflatoxin B1 (AFB1) mainly induces the transversion of G-->T in the third position of codon 249 of the p53 tumor suppressor gene, resulting in mutation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0450