Learn More
Aflatoxin B2-13C17 Solution, 0.5 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 1217470-98-8 |
| Flash Point | 2.0°C - Closed Cup |
| Linear Formula | 13C17H14O6 |
| MDL Number | MFCD16628324 |
| Synonym | Fully 13 C-labelled Aflatoxin B2 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 331.16 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 0.5 μg/mL in acetonitrile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.