Learn More
Enzo Life Sciences Aflatoxin B2 (1mg). CAS: 7220-81-7
SDP
Supplier:  Enzo Life Sciences ALX630103M001
Encompass_Preferred
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in methanol, methylene chloride (5mg/ml) or DMSO (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9625