Learn More
Cayman Chemical Aflatoxin G1-13C17, Purity: ≥98%, CAS: 1217444-07-9, 1.2 mL
SDP
Catalog No. 502991472
Encompass_Preferred
An internal standard for the quantification of aflatoxin G1 by GC- or LC-MS
Catalog No. 50-299-1472 Supplier Cayman Chemical Supplier No. 312621.2 ML
An internal standard for the quantification of aflatoxin G1 by GC- or LC-MS