Learn More
Enzo Life Sciences Aflatoxin G1 (1mg). CAS: 1165-39-5
SDP
Supplier:  Enzo Life Sciences ALX630104M001
Encompass_Preferred
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Purity: ≥98% (TLC). Appearance: White to yellow powder. Blue-green fluorescence. Solubility: Soluble in DMSO (10mg/ml), methylene chloride (5mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9627