Learn More
Enzo Life Sciences Aflatoxin G1 (5mg). CAS: 1165-39-5
SDP
Supplier:  Enzo Life Sciences ALX630104M005
Encompass_Preferred
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml), methylene chloride (5mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9628