Learn More
Cayman Chemical Aflatoxin G2-13C17, Purity: ≥98%, CAS: 1217462-49-1, 1.2 mL
SDP
Catalog No. 502991445
Encompass_Preferred
An internal standard for the quantification of aflatoxin G2 by GC- or LC-MS
Catalog No. 50-299-1445 Supplier Cayman Chemical Supplier No. 312631.2 ML
An internal standard for the quantification of aflatoxin G2 by GC- or LC-MS