Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin G2-13C17 Solution, 0.5 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111017925 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7925 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7925 Supplier MilliporeSigma Supelco Supplier No. 327771ML
Add to Cart
Add to Cart

Aflatoxin G2-13C17 is the 13C isotope-labeled analog of the mycotoxin, aflatoxin.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1217462-49-1
Flash Point 2.0°C - Closed Cup
Linear Formula 13C17H14O7
MDL Number MFCD14636316
Synonym Fully 13 C-labelled Aflatoxin G2
Type Analytical Standard
Quantity 1 mL
Formula Weight 347.16
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 0.5 μg/mL in acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.