Learn More
Cayman Chemical AflatoxIn G2 2.5mg
SDP
Supplier:  Cayman Chemical 112962.5 MG
Encompass_Preferred
A mycotoxin; lethal to ducklings (LD50 = 2.83 mg/kg); non-toxic to rats up to 200 mg/kg
Catalog No. 50-295-2933