Learn More
Enzo Life Sciences Aflatoxin G2 (5mg). CAS: 7241-98-7
SDP
Supplier:  Enzo Life Sciences ALX630106M005
Encompass_Preferred
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Purity: ≥98% (TLC). Solubility: Soluble in methylene chloride (5mg/ml), DMSO or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9632