Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin G2, crystalline, Thermo Scientific™

Catalog No. AC227370010
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227370010 1 mg
AC227370100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC227370010 Supplier Thermo Scientific Chemicals Supplier No. 227370010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7241-98-7
Molecular Formula C17H14O7
Molecular Weight (g/mol) 330.292
MDL Number MFCD00078141
InChI Key WPCVRWVBBXIRMA-WNWIJWBNSA-N
PubChem CID 2724362
ChEBI CHEBI:80705
SMILES COC1=C2C3=C(C(=O)OCC3)C(=O)OC2=C4C5CCOC5OC4=C1

Specifications

Color Yellow
Quantity 1 mg
Infrared Spectrum Authentic
Physical Form Powder
Beilstein 19, V,12, 79
Merck Index 15, 172
Synonym aflatoxin g2, dihydroaflatoxin g1, unii-2ms0d8wa29, ccris 4936, 1h,12h-furo 3',2':4,5 furo 2,3-h pyrano 3,4-c 1 benzopyran-1,12-dione, 3,4,7a,9,10,10a-hexahydro-5-methoxy-, 7ar,10as, 1h,12h-furo 3',2':4,5 furo 2,3-h pyrano 3,4-c 1 benzopyran-1,12-dione,3,4,7a,9,10,10a-hexahydro-5-methoxy-, 7ar,10as, 1h,12h-furo 3',2':4,5 furo 2,3-h pyrano 3,4-c 1 benzopyran-1,12-dione,3,4,7aalpha,9,10,10aalpha-hexahydro-5-methoxy, 3,4,7aalpha,9,10,10aalpha-hexahydro-5-methoxy-1h,12h-furo 3',2':4,5 furo 2,3-h pyrano 3,4-c 1-benzopyran-1,12-dione, aflatoxin g2 solution
Formula Weight 330.28
Percent Purity ≥98%
Chemical Name or Material Aflatoxin G2
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
May cause cancer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-643-4
RTECSNumber LV1700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.