Learn More
Enzo Life Sciences Aflatoxin M1 (0.1mg). CAS: 6795-23-9
SDP
Catalog No. 502009621
Encompass_Preferred
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Hydroxylated metabolite of aflatoxin B1. Maximum absorbance (λmax) at 228, 264 and 360nm. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in methylene chloride (1mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9621 Supplier Enzo Life Sciences Supplier No. ALX630095MC01
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Hydroxylated metabolite of aflatoxin B1. Maximum absorbance (λmax) at 228, 264 and 360nm. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in methylene chloride (1mg/ml). Long Term Storage: +4°C.