Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin Mix 4 Solution, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017102 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7102 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7102 Supplier MilliporeSigma Supelco Supplier No. 334152ML
Add to Cart
Add to Cart

Aflatoxins are a group of polyketide-derived furanocoumarins, which are known to contaminate the agricultural commodities. They pose a potential risk to livestock and human health because of their carcinogenic nature.

TRUSTED_SUSTAINABILITY

Specifications

Description B1, G1, B2, G2 in acetonitrile (20 μg/mL each)
MDL Number MFCD04974643
Quantity 2 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.