Learn More
New England Biolabs, Inc. AflII – 2000 units
SDP
Catalog No. 50812172
Encompass
A restriction endonuclease that recognizes the sequence C^TTAA_G.
Catalog No. 50-812-172 Supplier New England Biolabs, Inc. Supplier No. R0520S
A restriction endonuclease that recognizes the sequence C^TTAA_G.