Learn More
New England Biolabs, Inc. AflIII – 250 units
SDP
Catalog No. 50812213
Encompass
A restriction endonuclease that recognizes the sequence A^CRYG_T.
Catalog No. 50-812-213 Supplier New England Biolabs, Inc. Supplier No. R0541S
A restriction endonuclease that recognizes the sequence A^CRYG_T.