Learn More
Electron Microscopy Sciences Calibration Specimen 300-2D, Unmounted
SDP
Catalog No. 5031257
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. 300-2D, pattern: array of posts, nominal pitch: 300 nm, material: W-coated photoresist on Si.
Mount: Unmounted
Catalog No. 50-312-57 Supplier Electron Microscopy Sciences Supplier No. 801232D
Add to Cart
Add to Cart
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. 300-2D, pattern: array of posts, nominal pitch: 300 nm, material: W-coated photoresist on Si.
Mount: Unmounted