Learn More
Medchemexpress LLC AFM-30a hydrochloride | 99.8% | 502.97 | 1 ML
SDP
Catalog No. 5000438675
Encompass_Preferred
AFM-30a hydrochloride is a potent and selective protein arginine deiminase 2 (PAD2) inhibitor. It inhibits H3 citrullination and is intended for research into certain cancers and various autoimmune diseases, including rheumatoid arthritis (RA), multiple sclerosis, lupus, and ulcerative colitis.
  • Potent protein arginine deiminase 2 (PAD2) inhibitor.
  • Excellent PAD2-selectivity.
  • Inhibits H3 citrullination.
  • Can be used for research of certain cancers and autoimmune diseases.
  • Exhibited low cytotoxicity for cells in a cell viability assay.
  • Suppresses NLRP3 signaling and decreases airway remodeling.
Catalog No. 50-004-38675 Supplier Medchemexpress LLC Supplier No. HY125099A10MM/1ML
AFM-30a hydrochloride is a potent and selective protein arginine deiminase 2 (PAD2) inhibitor. It inhibits H3 citrullination and is intended for research into certain cancers and various autoimmune diseases, including rheumatoid arthritis (RA), multiple sclerosis, lupus, and ulcerative colitis.
  • Potent protein arginine deiminase 2 (PAD2) inhibitor.
  • Excellent PAD2-selectivity.
  • Inhibits H3 citrullination.
  • Can be used for research of certain cancers and autoimmune diseases.
  • Exhibited low cytotoxicity for cells in a cell viability assay.
  • Suppresses NLRP3 signaling and decreases airway remodeling.