Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afm-30a hydrochloride | 99.8% | 502.97 g/mol | C24H28ClFN6O3 | 5 MG
SDP

Catalog No. 5000264445
Encompass_Preferred

AFM-30a hydrochloride is the hydrochloride salt of AFM-30a, a potent and selective protein arginine deiminase 2 (PAD2) inhibitor for research applications. Supplied as a solid, the compound is characterized by high purity and includes manufacturer-recommended solubility and storage conditions for both solid and solution forms.

  • Potent PAD2 inhibitor with demonstrated selectivity.
  • High purity: 99.8%.
  • Hydrochloride salt to enhance solubility and handling.
  • Soluble in DMSO and water at ≥50 mg/mL for in vitro use.
  • Recommended storage: solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Supplied in small research quantities suitable for assay development.

Catalog No. 50-002-64445 Supplier Medchemexpress LLC Supplier No. HY125099A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AFM-30a hydrochloride is the hydrochloride salt of AFM-30a, a potent and selective protein arginine deiminase 2 (PAD2) inhibitor for research applications. Supplied as a solid, the compound is characterized by high purity and includes manufacturer-recommended solubility and storage conditions for both solid and solution forms.

  • Potent PAD2 inhibitor with demonstrated selectivity.
  • High purity: 99.8%.
  • Hydrochloride salt to enhance solubility and handling.
  • Soluble in DMSO and water at ≥50 mg/mL for in vitro use.
  • Recommended storage: solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Supplied in small research quantities suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.