Learn More
Aviva Systems Biology Corp AFM Antibody - middle region (ARP33804_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33804_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AFM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AFM. Alternate names: ALF, ALB2, ALBAApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-55152