Learn More
Electron Microscopy Sciences AFM Specimen Metal Discs | 20mm
SDP
Catalog No. 501927089
Encompass_Preferred
These are high quality metal discs with smooth edges and flat surfaces for use in Atomic force Microscopy.
Catalog No. 50-192-7089 Supplier Electron Microscopy Sciences Supplier No. 7501020
These are high quality metal discs with smooth edges and flat surfaces for use in Atomic force Microscopy.