Learn More
Electron Microscopy Sciences Calibration Specimen 750-HD, Unmounted
SDP
Catalog No. 5031253
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. AFM, SEM, product: 750-HD, pattern: array of flat bumps, nominal pitch: 750 (x), Z (100), material: Ni.
Mount: Unmounted
Catalog No. 50-312-53 Supplier Electron Microscopy Sciences Supplier No. 80124HD
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. AFM, SEM, product: 750-HD, pattern: array of flat bumps, nominal pitch: 750 (x), Z (100), material: Ni.
Mount: Unmounted