Learn More
Ted Pella Inc AFM SPECIMEN DISCS, 20MM 100/P
SDP
Catalog No. NC1955732
Encompass
High quality magnetic stainless steel (alloy 430) discs for Atomic Force Microscopy are offered, with smooth edges and consistently flat surfaces. The AFM/STM disks are 20 gauge which is 0.0359" (0.91mm) with a range of 0.0329" to 0.0389" (0.835mm to 0.988mm) thickness.
Catalog No. NC1955732 Supplier Ted Pella Inc Supplier No. 16219100PK
High quality magnetic stainless steel (alloy 430) discs for Atomic Force Microscopy are offered, with smooth edges and consistently flat surfaces. The AFM/STM disks are 20 gauge which is 0.0359" (0.91mm) with a range of 0.0329" to 0.0389" (0.835mm to 0.988mm) thickness.