Learn More
Electron Microscopy Sciences AFM/STM 90° Mount
SDP
Catalog No. 501930069
Encompass_Preferred
These mounts and sample holders make it easy to clamp and position cross sectional samples for AFM imaging. They're easy to attach to the magnetic sample holder of the AFM system. Compatible with most AFM systems on the market. Made from high quality magnetic stainless steel.
Catalog No. 50-193-0069 Supplier Electron Microscopy Sciences Supplier No. 7400001
These mounts and sample holders make it easy to clamp and position cross sectional samples for AFM imaging. They're easy to attach to the magnetic sample holder of the AFM system. Compatible with most AFM systems on the market. Made from high quality magnetic stainless steel.